Ansell™ Microchem™ 4000 Coveralls with Hood
Provides an exceptional barrier against organic and inorganic chemicals and biological agents. Ansell Microchem™ 4000 Coveralls with Hood is constructed of a unique multi-layer fabric known for its lightwieght, yet robust textile feel.
Overview
605.38 EUR valid until 2025-12-31
Use promo code "24380" to get your promotional price.
Additional Details:
Additional Details: Weight: 0.55000kg
Product Code. 15526444
Quantity Price
1 549.0  € / Pack of 10
Estimated Shipment
17-04-2026
Add to basket
  • Description and Specification
  • Alternative Products
Description and Specification
Specification
Product Type Protective Coveralls
Size 3X-Large
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color Green
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Double,Elastic
Cuff Style Double,Elastic
Neck Style Hooded
Hood Type 2-panel, Elastic
Closure Type Double Zipper Front Entry
Seam Type Taped,Welded
Seam Type Taped,Welded
Resistance Chemicals
Certifications/Compliance Type 3/4 EN14605, Type 5 EN ISO 13982-1, EN14126, EN1073-2, EN1149-5
No. of Pockets None
Style Includes Hood
  • Coverall with elasticated 2-piece hood and front entry double-zip system.
  • Elasticated double cuffs (knitted inner cuff), waist and ankles.
  • Permeation tested against over 180 chemicals, including chemical warfare agents
  • Textile-like inner improves wearer acceptance
  • Anti-static according to EN 1149-5
Alternative Products