Code:
NEW
Sodium sulfate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., anhydrous, ≥99.0%, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., anhydrous,≥99.0%
Additional Details:
CAS Number:
7757-82-6
Weight:
2.50000kg
| Quantity | 2.5 kg |
|---|
Chemical Identifiers
| CAS | 7757-82-6 |
|---|---|
| Molecular Formula | Na2O4S |
| Molecular Weight (g/mol) | 142.04 |
| MDL Number | MFCD00003504 |
| InChI Key | PMZURENOXWZQFD-UHFFFAOYSA-L |
| Synonym | sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 |
| PubChem CID | 24436 |
| ChEBI | CHEBI:32149 |
| IUPAC Name | disodium;sulfate |
| SMILES | [Na+].[Na+].[O-]S([O-])(=O)=O |
Specification
| Chemical Name or Material | Sodium Sulfate |
|---|---|
| Name Note | Reag. Ph. Eur. reag. ISO |
| CAS | 7757-82-6 |
| Physical Form | Anhydrous |
| Quantity | 2.5 kg |
| Molecular Formula | Na2O4S |
| Linear Formula | Na2SO4 |
| MDL Number | MFCD00003504 |
| UN Number | NONH for all modes of transport |
| Synonym | sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 |
| Synonym | sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 |
| Synonym | sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 |
| Synonym | sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 |
| Synonym | sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 |
| Synonym | sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 |
| Synonym | sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 |
| Synonym | sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 |
| Synonym | sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 |
| Synonym | sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 |
| InChI Key | PMZURENOXWZQFD-UHFFFAOYSA-L |
| SMILES | [Na+].[Na+].[O-]S([O-])(=O)=O |
| IUPAC Name | disodium;sulfate |
| Molecular Weight (g/mol) | 142.04 |
| PubChem CID | 24436 |
| ChEBI | CHEBI:32149 |
| Formula Weight | 142.04g/mol |
| Percent Purity | ≥99% |
| Grade | ACS/puriss. p.a. |
| Density | 2.68g/mL at 25°C |
Technical documents
[Sodium sulfate]
Product Identifier
- Sodium sulfate