344.28 EUR valid until 2025-07-31
Use promo code "24075" to get your promotional price.
Use promo code "24075" to get your promotional price.
Autoclavable | Autoclavable |
---|---|
Color | Dark Gray |
For Use With | Pipette Tip 2.5 μL |
Number of Channels | 1 |
Product Line | Reference 2 |
Type | Pipette |
Volume (Metric) | 0.1 to 2.5 μL |
For Use With (Equipment) | 2.5 μL pipette tips |
Includes | epT.I.P.S.™ Box 2.0 with 96 Tips |
Increments | 0.002 μL |
Color Code | Dark Gray |