Use promo code "25339" to get your promotional price.
Thermo Scientific Acros Ethyl acetate, 99.5%, for analysis
Use promo code "25339" to get your promotional price.
Quantity | 1 L |
---|---|
Packaging | Glass Bottle |
Chemical Identifiers
CAS | 141-78-6 |
---|---|
Molecular Formula | C4H8O2 |
Molecular Weight (g/mol) | 88.106 |
MDL Number | MFCD00009171 |
InChI Key | XEKOWRVHYACXOJ-UHFFFAOYSA-N |
Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
PubChem CID | 8857 |
ChEBI | CHEBI:27750 |
IUPAC Name | ethyl acetate |
SMILES | CCOC(=O)C |
Specification
Chemical Name or Material | Ethyl acetate |
---|---|
Melting Point | -83.5°C |
CAS | 141-78-6 |
Density | 0.9020g/mL |
Boiling Point | 75.0°C to 78.0°C |
Quantity | 1 L |
Flash Point | −4°C |
Packaging | Glass Bottle |
Physical Form | Liquid |
Refractive Index | 1.3710 to 1.3740 |
Assay Percent Range | 99.5% |
Residue after Evaporation | 0.001% max. |
Specific Gravity | 0.902 |
Molecular Formula | C4H8O2 |
Linear Formula | CH3CO2C2H5 |
MDL Number | MFCD00009171 |
Viscosity | 0.45 mPa.s (20°C) |
Beilstein | 02, 125 |
Water | 0.05% max. |
Merck Index | 15, 3811 |
Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
Synonym | ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester |
Solubility Information | Solubility in water: 80g/L (20°C). Other solubilities: soluble in benzene, acetone, chloroform, alcohol,, ether |
InChI Key | XEKOWRVHYACXOJ-UHFFFAOYSA-N |
SMILES | CCOC(=O)C |
IUPAC Name | ethyl acetate |
Molecular Weight (g/mol) | 88.106 |
PubChem CID | 8857 |
ChEBI | CHEBI:27750 |
Formula Weight | 88.11 |
Percent Purity | 99.5% |
Grade | Analytical |
Technical documents
[Ethyl acetate]
[Danger]
[{code=FLAM. LIQ. 2, sequence=1, description=Flammable liquid Category 2}, {code=EYE DAM. 2, sequence=2, description=Serious eye damage/eye irritation Category 2}, {code=STOT SE 3, sequence=3, description=Specific target organ toxicity Category 3}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H319, sequence=2, description=Causes serious eye irritation.}, {code=H336, sequence=3, description=May cause drowsiness or dizziness.}, {code=EUH066, sequence=91, description=Repeated exposure may cause skin dryness or cracking.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P240, sequence=52, description=Ground/bond container and receiving equipment.}, {code=P261, sequence=53, description=Avoid breathing dust/fume/gas/mist/vapours/spray.}, {code=P280, sequence=54, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- Ethyl acetate
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H319- Causes serious eye irritation.
- H336- May cause drowsiness or dizziness.
- EUH066- Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240- Ground/bond container and receiving equipment.
- P261- Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.