Thermo Scientific Isopropanol, 99.5+%, extra pure
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Overview
190.00 GBP valid until 2025-12-31
Use promo code "24348" to get your promotional price.
Code: 29
Additional Details:
Additional Details: CAS Number: 67-63-0 Weight: 7.85000kg Transport: UN number : 1219 Chem class : 3 Pack group : II
Quantity 10 L
Packaging Plastic Drum
Disclaimers: RUO – Research Use Only
Product Code. 10085103
Quantity Price
1 £ 207.0 / 10 L
Estimated Shipment
31-07-2025
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
  • Special Offers
Description and Specification
Chemical Identifiers
CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O
Specification
Chemical Name or Material Isopropanol
Melting Point -89.5°C
CAS 67-63-0
Density 0.7850g/mL
Boiling Point 81.0°C to 83.0°C
Quantity 10 L
Flash Point 12°C
Packaging Plastic Drum
Physical Form Liquid
Refractive Index 1.3760 to 1.3780
Assay Percent Range 99.5+%
Specific Gravity 0.785
Molecular Formula C3H8O
Linear Formula (CH3)2CHOH
MDL Number MFCD00011674
Viscosity 2.27 mPa.s (20°C)
Water 0.2% max.
Merck Index 15, 5254
Synonym isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
Synonym isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
Synonym isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
Synonym isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
Synonym isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
Synonym isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
Synonym isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
Synonym isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
Synonym isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
Synonym isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
Solubility Information Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, methylene chloride,, dmf and acetone, miscible with alcohol, ether and chloroform, insoluble in salts solutions
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES CC(C)O
IUPAC Name propan-2-ol
Molecular Weight (g/mol) 60.096
PubChem CID 3776
ChEBI CHEBI:17824
Formula Weight 60.1
Percent Purity ≥99.5%
Grade Extra Pure
Alternative Products
Documentation
Technical documents
Safety and Handling
[Isopropanol]
[Danger]
[{code=EYE DAM. 2, sequence=1, description=Serious eye damage/eye irritation Category 2}, {code=FLAM. LIQ. 2, sequence=2, description=Flammable liquid Category 2}, {code=STOT SE 3, sequence=3, description=Specific target organ toxicity Category 3}]
[{code=H319, sequence=1, description=Causes serious eye irritation.}, {code=H336, sequence=2, description=May cause drowsiness or dizziness.}, {code=H225, sequence=3, description=Highly flammable liquid and vapour.}]
[{code=P210, sequence=50, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P240, sequence=51, description=Ground/bond container and receiving equipment.}, {code=P261, sequence=52, description=Avoid breathing dust/fume/gas/mist/vapours/spray.}, {code=P280, sequence=53, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
  • Isopropanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319- Causes serious eye irritation.
  • H336- May cause drowsiness or dizziness.
  • H225- Highly flammable liquid and vapour.
Precautionary Statement
  • P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240- Ground/bond container and receiving equipment.
  • P261- Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Special Offers