Azlon Azlon™ HDPE, Jerry Can
Capacity Metric: 5 L
Overview
12.47 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: A0
Additional Details:
Additional Details: Weight: 0.50000kg
Product Code. 10115674
Quantity Price
1 £ 21.15 / Each
Estimated Shipment
26-09-2025
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Specification
Capacity (English) 1.3 gal.
Capacity (Metric) 5 L
Dimensions (L x W x H) 186 x 134 x 282mm
Material HDPE
No. per Pack 1 Ea.
Product Type Jerry Can
  • Lightweight and robust
  • Corrosion resistant
  • Suitable for food contact applications in accordance with EU and U.S. FDA regulations