Use promo code "24117" to get your promotional price.
Invitrogen™ RNaseZap™ RNase Decontamination Solution
RNaseZap™ RNase Decontamination Solution is a surface decontamination solution that destroys RNases on contact. You simply spray RNaseZap™ Solution onto the surface to be decontaminated and rinse it off with RNase-free water.
792.30 GBP valid until 2025-08-24
Use promo code "24117" to get your promotional price.
Use promo code "24117" to get your promotional price.
Code:
50
Additional Details:
Weight:
4.46000kg
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specification
Product Type | RNase |
---|---|
Content And Storage | Store at room temperature. |
Shipping Condition | Room Temperature |
Quantity | 4 L |
Reagent Type | RNase Control Reagent |
Product Line | Ambion™,RNaseZap™ |
Product Line | Ambion™,RNaseZap™ |