RETSCH Stainless Steel Grinding Balls
Designed for use with Retsch™ mixer mills. Retsch™ Stainless Steel Grinding Balls are ideal for disruption of biological cells as well as for DNA/RNA extraction.
Overview
97.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Additional Details:
Additional Details: Weight: 0.02000kg
Product Code. 10196541
Quantity Price
1 £ 101.0 / Each
Estimated Shipment
06-10-2025
Add to basket
  • Description and Specification
  • Alternative Products
Description and Specification
Specification
Product Type Grinding Balls
Diameter (Metric) 3 mm
For Use With (Equipment) Reaction Vials
Material Stainless Steel
  • For use with MM 200 and MM 400 Mixer Mills
Alternative Products