| CAS |
107-21-1
|
| Melting Point |
-13°C
|
| Color |
Colorless
|
| pH |
5.5 to 7.5
|
| Boiling Point |
196°C
|
| Packaging |
Amber glass bottle
|
| Molecular Formula |
C2H6O2
|
| MDL Number |
2885
|
| Quantity |
2.5 L
|
| Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
| Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
| Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
| Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
| Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
| Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
| Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
| Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
| Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
| Synonym |
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
|
| InChI Key |
LYCAIKOWRPUZTN-UHFFFAOYSA-N
|
| SMILES |
C(CO)O
|
| IUPAC Name |
ethane-1,2-diol
|
| Molecular Weight (g/mol) |
62.068
|
| PubChem CID |
174
|
| Vapor Density |
2.14
|
| ChEBI |
CHEBI:30742
|
| Formula Weight |
62.06g/mol
|
| Vapor Pressure |
0.12mmHg at 20°C
|
| Physical Form |
Viscous Liquid
|