275.02 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Use promo code "24135" to get your promotional price.
Connectors | Hose |
---|---|
Disposable | Single-use |
For Use With (Equipment) | 4.5 cm bottle neck |
Material | PES |
Material (Membrane) | PES |
Pore Size | 0.22 μm |
Sterility | Sterile |
Filtration Area | 13.6 cm2 |
For Use With (Application) | Ideal for filtration of tissue culture media and components, biological fluids, other solutions |
Graduated | Yes |
Includes | Square membrane filter, graduated funnel of clear polystyrene with polyethylene neck yellow adapter, and tubing adapter |
No. per Case | 48/Cs. |
No. per Pack | 1/Pk. |
Type | Bottle Top Vacuum Filter |
Volume (Metric) | 150 mL |
Diameter (Metric) | 5 cm |