Code:
66
Corning™ Costar™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Sterile and nonpyrogenic microplates with low evaporation lids.
Additional Details:
Weight:
3.31200kg
Specification
Format | Standard |
---|---|
Lid | With Lid |
Culture Area | 0.32 cm2 |
Array | 8 x 12 |
Packaging | Bulk, 10 Bags x 5 Plates |
Binding Type | TC-Treated |
Color | Clear |
Material | Polystyrene |
No. of Wells | 96 |
Volume (Metric) Working | 0.1 to 0.2 mL |
Sterility | Sterile |
Surface Treatment | Cell Culture |
Total Volume (Metric) Well | 0.36 mL |
Well Shape | Flat Bottom (F-well) |
No. per Pack | 50 |
Quantity | 50/Cs. |
Diameter (Metric) Well | 0.64 cm |