36.64 GBP valid until 2025-12-31
Use promo code "24361" to get your promotional price.
Use promo code "24361" to get your promotional price.
| Quantity | 20 mL |
|---|---|
| Packaging | Poly Bottle |
| CAS | 1397-89-3 |
|---|---|
| Molecular Formula | C47H73NO17 |
| Molecular Weight (g/mol) | 924.09 |
| MDL Number | MFCD00877763 |
| InChI Key | APKFDSVGJQXUKY-ZNVUZQDLSA-N |
| PubChem CID | 134129663 |
| IUPAC Name | (1S,3R,18S,19R,20R,21S,25R,27R,30R,31R,33S,35R,37S,38R)-3-[(2R,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,30,31,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,12,14,16-heptaene-38-car |
| SMILES | C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O |
| Chemical Name or Material | Amphotericin B |
|---|---|
| Type | Amphotericin B |
| Name Note | 250μg/mL |
| Physical Form | Liquid |
| CAS | 1397-89-3 |
| Packaging | Poly Bottle |
| Sterility | Filter Sterilized |
| Color | Yellow |
| Quantity | 20 mL |
| Molecular Formula | C47H73NO17 |
| MDL Number | MFCD00877763 |
| Boiling Point | 100°C |
| Merck Index | 15, 582 |
| InChI Key | APKFDSVGJQXUKY-ZNVUZQDLSA-N |
| SMILES | C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O |
| IUPAC Name | (1S,3R,18S,19R,20R,21S,25R,27R,30R,31R,33S,35R,37S,38R)-3-[(2R,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,30,31,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,12,14,16-heptaene-38-car |
| Molecular Weight (g/mol) | 924.09 |
| PubChem CID | 134129663 |
| Formula Weight | 924.1 |