Greiner Bio-One Polystyrene Petri Dishes
Manufactured from highly transparent polystyrene for its heat resistant properties to enable use with hot agar. Greiner BioOne™ Polystyrene Petri Dishes are available in a wide variety of formats and sizes to accommodate multiple research needs.
Additional Details:
Weight:
6.84900kg
Specification
Capacity (Metric) | 80 mL |
---|---|
Disposable | Single-use |
Material | Polystyrene |
Shape | Round |
Sterility | Non-sterile |
Quantity | 480 Pack |
Vent | Yes |
Dimensions (Dia. x H) | 94 x 16 mm |
Height (Metric) | 16 mm |
Diameter (Metric) | 94 mm |