Use promo code "25339" to get your promotional price.
Potassium fluoride, 99%, extra pure, anhydrous, Thermo Scientific Chemicals
Use promo code "25339" to get your promotional price.
| Quantity | 500 g |
|---|---|
| Packaging | Plastic bottle |
Chemical Identifiers
| CAS | 7789-23-3 |
|---|---|
| Molecular Formula | FK |
| Molecular Weight (g/mol) | 58.10 |
| MDL Number | MFCD00011398 |
| InChI Key | NROKBHXJSPEDAR-UHFFFAOYSA-M |
| Synonym | potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 |
| PubChem CID | 522689 |
| ChEBI | CHEBI:66872 |
| SMILES | [F-].[K+] |
Specification
| Boiling Point | 1505.0°C |
|---|---|
| Chemical Name or Material | Potassium fluoride |
| Melting Point | 860.0°C |
| Name Note | 99% |
| CAS | 7789-23-3 |
| Color | White |
| Physical Form | Crystalline Powder, Crystals and/or Chunks |
| Quantity | 500 g |
| Assay Percent Range | 99% |
| Molecular Formula | FK |
| Linear Formula | KF |
| MDL Number | MFCD00011398 |
| Fieser | 01,933; 02,346; 05,555; 06,481; 08,410; 10,325; 11,434; 12,406; 13,256; 15,167; 16,282 |
| Merck Index | 15, 7753 |
| Synonym | potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 |
| Synonym | potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 |
| Synonym | potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 |
| Synonym | potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 |
| Synonym | potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 |
| Synonym | potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 |
| Synonym | potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 |
| Synonym | potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 |
| Synonym | potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 |
| Synonym | potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 |
| Solubility Information | Solubility in water: 920g/L (18°C). Other solubilities: soluble in hydrofluoric acid, insoluble in alcohol unless water is present |
| InChI Key | NROKBHXJSPEDAR-UHFFFAOYSA-M |
| SMILES | [F-].[K+] |
| IUPAC Name | potassium;fluoride |
| Molecular Weight (g/mol) | 58.10 |
| PubChem CID | 522689 |
| ChEBI | CHEBI:66872 |
| Formula Weight | 58.09 |
| Percent Purity | 99% |
| Grade | Extra Pure |
| Identification | Pass Test |
| Packaging | Plastic bottle |
| Density | 2.48 |
Technical documents
[Potassium fluoride]
[Danger]
[{code=ACUTE TOX. 3, sequence=1, description=Acute toxicity Category 3}, {code=EYE DAM. 1, sequence=2, description=Serious eye damage/eye irritation Category 1}]
[{code=H301, sequence=1, description=Toxic if swallowed.}, {code=H311, sequence=2, description=Toxic in contact with skin.}, {code=H331, sequence=3, description=Toxic if inhaled.}, {code=H318, sequence=4, description=Causes serious eye damage.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=52, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P302+P352, sequence=53, description=IF ON SKIN: Wash with plenty of water/.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=55, description=Immediately call a POISON CENTER/doctor/.}]
[{code=GHS06, sequence=1, description=SKULL & CROSSBONES}, {code=GHS05, sequence=2, description=CORROSION}]
Product Identifier
- Potassium fluoride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H301- Toxic if swallowed.
- H311- Toxic in contact with skin.
- H331- Toxic if inhaled.
- H318- Causes serious eye damage.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352- IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.