Code:
66
Corning™ Roller Bottle Caps
Easy-grip polyethylene
Additional Details:
Weight:
0.06207kg
Specification
| Color | Red |
|---|---|
| Material | Polyethylene |
| No. per Case | 300/Cs. |
| No. per Pack | 1/Pk. |
| Sterility | Sterile |
| Thread Size (Metric) | 31 mm |
| Neck Finish | 31 mm |
| For Use With (Equipment) | Roller bottle |