Sodium thiosulfate, 98.5%, extra pure, anhydrous
CAS: 7772-98-7 | Na2O3S2 | 158.10 g/mol
Overview
93.93 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: 29
Additional Details:
Additional Details: CAS Number: 7772-98-7 Weight: 2.50000kg
Quantity 2.5 kg
Packaging Plastic bottle
Disclaimers: RUO – Research Use Only
Product Code. 10472011
Quantity Price
1 £ 119.0 / 2.50kg
Estimated Shipment
07-07-2026
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Chemical Identifiers
CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
PubChem CID 24477
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O
Specification
Chemical Name or Material Sodium thiosulfate
Melting Point 48.0°C
CAS 7772-98-7
Color White
pH 6.0 to 8.5 (5% soln. at 20°C)
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 98% min. (on dry substance) (Iodimetry)
Molecular Formula Na2O3S2
Linear Formula Na2S2O3
MDL Number MFCD00003499
Merck Index 15,8821
Synonym sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
Synonym sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
Synonym sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
Synonym sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
Synonym sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
Synonym sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
Synonym sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
Synonym sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
Synonym sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
Synonym sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
Solubility Information Solubility in water: 700g/L (20°C). Other solubilities: practically insoluble in alcohol
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O
IUPAC Name disodium sulfanidesulfonate
Molecular Weight (g/mol) 158.10
PubChem CID 24477
Formula Weight 158.1
Percent Purity 98.5%
Grade Extra Pure
Loss on Drying 2% (105°C, 2 hrs) max.
Packaging Plastic bottle
Reduction reagent