| Chemical Name or Material |
Sodium bicarbonate
|
| Melting Point |
270.0°C
|
| CAS |
144-55-8
|
| Color |
White
|
| Physical Form |
Fine Crystalline Powder
|
| Quantity |
25 kg
|
| Assay Percent Range |
99+%
|
| Molecular Formula |
CHNaO3
|
| Linear Formula |
NaHCO3
|
| MDL Number |
MFCD00003528
|
| Fieser |
03,260; 05,595; 11,476; 17,180
|
| Merck Index |
15, 8719
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Solubility Information |
Solubility in water: 9g/100mL (20°C). Other solubilities: 16.4g/100mL water (60°C),insoluble in alcohol
|
| InChI Key |
UIIMBOGNXHQVGW-UHFFFAOYSA-M
|
| SMILES |
[Na+].OC([O-])=O
|
| IUPAC Name |
sodium;hydrogen carbonate
|
| Molecular Weight (g/mol) |
84.01
|
| PubChem CID |
516892
|
| ChEBI |
CHEBI:32139
|
| Formula Weight |
84.01
|
| Percent Purity |
≥99%
|
| Grade |
Extra Pure
|
| Packaging |
Fibre drum
|