Macherey-Nagel™ Nucleosil™ 100-5 Analytical HPLC Standard EC Column
Column Format: Analytical Column; Carbon Load: 25%; Pore Size: 100 Å; Column Phase: Reversed Phase
Overview
Code: 16
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 10506671
Quantity Price
1 £ 494.0 / Each
In stock, ready to ship: 1
Add to basket
  • Description and Specification
  • Alternative Products
Description and Specification
Specification
Column Chemistry Silica
Column Format Analytical Column
Crosslinking Polymeric octadecyl
Diameter (Metric) 4.6 mm
For Use With HPLC
Length (Metric) 150 mm
Max. Pressure 5800 psi (400 bar)
Mobile Phase Compatibility Acetonitrile/water
Particle Size 5 μm
pH 1 to 9
Pore Size 100 Å
Quantity 1 Ea.
Solvents Acetonitrile/water
Surface Area 350 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18 AB
Packing Material Silica
Carbon Load 25%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Type Analytical EC Column
Alternative Products