129.84 GBP valid until 2025-08-31
Use promo code "24124" to get your promotional price.
Use promo code "24124" to get your promotional price.
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
---|---|
Format | Standard |
Lid | Without Lid |
Array | 8 x 12 |
Packaging | 80 Cases |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
No. of Wells | 96 |
Sterility | Sterile |
Surface Treatment | Non-treated |
Well Shape | Conical Bottom (V-well) |
No. per Case | 80 |
No. per Pack | 10 |
Quantity | 80/Cs. |