44.85 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Sterility | Non-sterile |
---|---|
Type | Cuvette |
Color | Clear |
Disposable | Single-use |
Capacity (Metric) | 70 to 850 μL |
Material | Plastic |
Includes | Cuvette Only |
Quantity | 100 Pack, Individually Wrapped |
Path Length | 10 mm |
Wavelength Range | 230 to 900 nm |
Height (Metric) | 45 mm |
For Use With (Equipment) | Spectrophotometers and photometers |
Width (Metric) | 12.5 mm |
For Use With (Application) | DNA, RNA, Protein Analysis, Molecular Biology |
Lightpath | 10 mm |
Shape | Rectangle |
Manufacturing Quality | CERTIFIED LIFE SCIENCE QUALITY |