Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom Microplate
Color:Black; Sterility:Sterile; Lid:With Lid
Additional Details:
Weight:
0.80000kg
Specification
Format | Standard |
---|---|
Lid | With Lid |
Array | 8 x 12 |
Autoclavable | Not Autoclavable |
Color | Black |
Material | Polystyrene |
No. of Wells | 96 |
Volume (Metric) Working | 25 to 340 μL |
Sterility | Sterile |
Surface Treatment | Cell Culture |
Total Volume (Metric) Well | 392 μL |
Well Shape | Flat Bottom (F-well) |
Max. RCF | 4800 x g (swinging bucket rotor) |
No. per Case | 32 |
No. per Pack | 8 |
Quantity | 32/Cs. |
Temperature Range (Metric) | -20°C to +60°C |
Well Design | Chimney |