| Chemical Name or Material |
Sodium hydrogen carbonate
|
| Name Note |
For ACS analysis
|
| CAS |
144-55-8
|
| Color |
White
|
| Physical Form |
Granules
|
| Quantity |
2 kg
|
| Assay Percent Range |
99.7-100.3%
|
| Molecular Formula |
CHNaO3
|
| MDL Number |
MFCD00003528
|
| Beilstein |
4153970
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Synonym |
sodium bicarbonate,sodium hydrogen carbonate,baking soda,carbonic acid monosodium salt,sodium acid carbonate,bicarbonate of soda,sodium hydrogencarbonate,meylon,acidosan,neut
|
| Solubility Information |
Soluble in water and methanol. Insoluble in ethanol.
|
| InChI Key |
UIIMBOGNXHQVGW-UHFFFAOYSA-M
|
| SMILES |
[Na+].OC([O-])=O
|
| IUPAC Name |
sodium;hydrogen carbonate
|
| Molecular Weight (g/mol) |
84.01
|
| PubChem CID |
516892
|
| ChEBI |
CHEBI:32139
|
| Formula Weight |
84.01
|
| Grade |
ACS
|
| Odor |
Odorless
|
| Refractive Index |
1.5
|
| Density |
2.159
|