Use promo code "25339" to get your promotional price.
Potassium tert-butoxide, 97%
Use promo code "25339" to get your promotional price.
| Quantity | 250 g |
|---|
Chemical Identifiers
| CAS | 865-47-4 |
|---|---|
| Molecular Formula | C4H9KO |
| Molecular Weight (g/mol) | 112.213 |
| MDL Number | MFCD00012162 |
| InChI Key | LPNYRYFBWFDTMA-UHFFFAOYSA-N |
| Synonym | potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok |
| PubChem CID | 23665647 |
| IUPAC Name | potassium;2-methylpropan-2-olate |
| SMILES | CC(C)(C)[O-].[K+] |
Specification
| Chemical Name or Material | Potassium tert-butoxide |
|---|---|
| CAS | 865-47-4 |
| Melting Point | ∼255°C (decomposition) |
| Molecular Formula | C4H9KO |
| Linear Formula | (CH3)3COK |
| MDL Number | MFCD00012162 |
| Quantity | 250 g |
| UN Number | UN3206 |
| Beilstein | 3556712 |
| Sensitivity | Air and moisture sensitive |
| Synonym | potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok |
| Synonym | potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok |
| Synonym | potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok |
| Synonym | potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok |
| Synonym | potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok |
| Synonym | potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok |
| Synonym | potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok |
| Synonym | potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok |
| Synonym | potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok |
| Synonym | potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok |
| Solubility Information | Soluble in hexane,toluene,diethyl ether and terahydrofuran. |
| InChI Key | LPNYRYFBWFDTMA-UHFFFAOYSA-N |
| SMILES | CC(C)(C)[O-].[K+] |
| IUPAC Name | potassium;2-methylpropan-2-olate |
| Molecular Weight (g/mol) | 112.213 |
| PubChem CID | 23665647 |
| Formula Weight | 112.22 |
| Percent Purity | 97% |
Technical documents
[Potassium tert-butoxide]
[Danger]
[{code=H228, sequence=1, description=Flammable solid.}, {code=H252, sequence=2, description=Self-heating in large quantities; may catch fire.}, {code=H314, sequence=3, description=Causes severe skin burns and eye damage.}, {code=H318, sequence=4, description=Causes serious eye damage.}, {code=EUH014, sequence=91, description=Reacts violently with water.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=53, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=55, description=Immediately call a POISON CENTER/doctor/.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: Potassium tert-butoxide}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS05, sequence=2, description=CORROSION}]
Product Identifier
- Potassium tert-butoxide
Signal Word
- Danger
Hazard Statement
- H228- Flammable solid.
- H252- Self-heating in large quantities; may catch fire.
- H314- Causes severe skin burns and eye damage.
- H318- Causes serious eye damage.
- EUH014- Reacts violently with water.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
Supplemental Information
- MIXTURE LIST- Contain: Potassium tert-butoxide