Oxalyl chloride, 98%
CAS: 79-37-8 | C2Cl2O2 | 126.92 g/mol
Overview
54.10 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: CAS Number: 79-37-8 Weight: 0.10000kg Transport: UN number : 3130 Chem class : 4.3 Pack group : I
Quantity 100 g
Disclaimers: RUO – Research Use Only
Product Code. 11478700
Quantity Price
1 £ 54.7 / 100g
Estimated Shipment
17-09-2026
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
Description and Specification
Chemical Identifiers
CAS 79-37-8
Molecular Formula C2Cl2O2
Molecular Weight (g/mol) 126.92
MDL Number MFCD00000704
InChI Key CTSLXHKWHWQRSH-UHFFFAOYSA-N
Synonym oxalyl chloride,ethanedioyl dichloride,oxalic dichloride,oxaloyl chloride,oxalic acid dichloride,oxalic acid chloride,ethanedioyl chloride,oxalylchloride,unii-r4y96317dw,cocl 2
PubChem CID 65578
IUPAC Name oxalyl dichloride
SMILES C(=O)(C(=O)Cl)Cl
Specification
Chemical Name or Material Oxalyl chloride
CAS 79-37-8
Melting Point -12°C
Density 1.455
Boiling Point 63°C to 64°C
Assay Percent Range 0.98
Odor Acrid
Molecular Formula C2Cl2O2
Refractive Index 1.429
MDL Number MFCD00000704
Quantity 100 g
UN Number UN2927
Beilstein 1361988
Sensitivity Moisture sensitive
Merck Index 14,6914
Synonym oxalyl chloride,ethanedioyl dichloride,oxalic dichloride,oxaloyl chloride,oxalic acid dichloride,oxalic acid chloride,ethanedioyl chloride,oxalylchloride,unii-r4y96317dw,cocl 2
Synonym oxalyl chloride,ethanedioyl dichloride,oxalic dichloride,oxaloyl chloride,oxalic acid dichloride,oxalic acid chloride,ethanedioyl chloride,oxalylchloride,unii-r4y96317dw,cocl 2
Synonym oxalyl chloride,ethanedioyl dichloride,oxalic dichloride,oxaloyl chloride,oxalic acid dichloride,oxalic acid chloride,ethanedioyl chloride,oxalylchloride,unii-r4y96317dw,cocl 2
Synonym oxalyl chloride,ethanedioyl dichloride,oxalic dichloride,oxaloyl chloride,oxalic acid dichloride,oxalic acid chloride,ethanedioyl chloride,oxalylchloride,unii-r4y96317dw,cocl 2
Synonym oxalyl chloride,ethanedioyl dichloride,oxalic dichloride,oxaloyl chloride,oxalic acid dichloride,oxalic acid chloride,ethanedioyl chloride,oxalylchloride,unii-r4y96317dw,cocl 2
Synonym oxalyl chloride,ethanedioyl dichloride,oxalic dichloride,oxaloyl chloride,oxalic acid dichloride,oxalic acid chloride,ethanedioyl chloride,oxalylchloride,unii-r4y96317dw,cocl 2
Synonym oxalyl chloride,ethanedioyl dichloride,oxalic dichloride,oxaloyl chloride,oxalic acid dichloride,oxalic acid chloride,ethanedioyl chloride,oxalylchloride,unii-r4y96317dw,cocl 2
Synonym oxalyl chloride,ethanedioyl dichloride,oxalic dichloride,oxaloyl chloride,oxalic acid dichloride,oxalic acid chloride,ethanedioyl chloride,oxalylchloride,unii-r4y96317dw,cocl 2
Synonym oxalyl chloride,ethanedioyl dichloride,oxalic dichloride,oxaloyl chloride,oxalic acid dichloride,oxalic acid chloride,ethanedioyl chloride,oxalylchloride,unii-r4y96317dw,cocl 2
Synonym oxalyl chloride,ethanedioyl dichloride,oxalic dichloride,oxaloyl chloride,oxalic acid dichloride,oxalic acid chloride,ethanedioyl chloride,oxalylchloride,unii-r4y96317dw,cocl 2
Solubility Information Oxalyl chloride Slightly miscible with water.Miscible with ether,benzene and chloroform.
InChI Key CTSLXHKWHWQRSH-UHFFFAOYSA-N
SMILES C(=O)(C(=O)Cl)Cl
IUPAC Name oxalyl dichloride
Molecular Weight (g/mol) 126.92
PubChem CID 65578
Formula Weight 126.93
Percent Purity 98%
Physical Form Liquid
Applications
Oxalyl chloride is mainly used as a catalyst in the oxidation of alcohols to the corresponding aldehydes and ketones. It is actively used for the synthesis of acid chlorides from acids. In the Fridel-Crafts acylation reaction, it reacts with aromatic compounds to get the corresponding acyl choride using aluminum chloride as catalyst. Further, it is utilized to prepare dioxane tetrketone, an oxide of carbon.
Solubility
Oxalyl chloride Slightly miscible with water.Miscible with ether, benzene and chloroform.
Notes
Moisture sensitive. Incompatible with bases, alcohols, water, amines and metals.
Alternative Products
Documentation
Technical documents
Safety and Handling
[Oxalyl chloride, 98%]
[Danger]
[{code=H260, sequence=1, description=In contact with water releases flammable gases which may ignite spontaneously.}, {code=H301, sequence=2, description=Toxic if swallowed.}, {code=H314, sequence=3, description=Causes severe skin burns and eye damage.}, {code=H318, sequence=4, description=Causes serious eye damage.}, {code=H331, sequence=5, description=Toxic if inhaled.}]
[{code=P231, sequence=51, description=Handle under inert gas.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=53, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=54, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P304+P340, sequence=55, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=56, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=57, description=Immediately call a POISON CENTER/doctor/.}, {code=P335+P334, sequence=58, description=Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: ethanedioyl dichloride}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS05, sequence=2, description=CORROSION}, {code=GHS06, sequence=3, description=SKULL & CROSSBONES}]
Product Identifier
  • Oxalyl chloride, 98%
Signal Word
  • Danger
Hazard Statement
  • H260- In contact with water releases flammable gases which may ignite spontaneously.
  • H301- Toxic if swallowed.
  • H314- Causes severe skin burns and eye damage.
  • H318- Causes serious eye damage.
  • H331- Toxic if inhaled.
Precautionary Statement
  • P231- Handle under inert gas.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310- Immediately call a POISON CENTER/doctor/.
  • P335+P334- Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
Supplemental Information
  • MIXTURE LIST- Contain: ethanedioyl dichloride