Use promo code "24128" to get your promotional price.
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%
Use promo code "24128" to get your promotional price.
| Quantity | 100 g |
|---|
Chemical Identifiers
| CAS | 6674-22-2 |
|---|---|
| Molecular Formula | C9H16N2 |
| Molecular Weight (g/mol) | 152.241 |
| MDL Number | MFCD00006930 |
| InChI Key | GQHTUMJGOHRCHB-UHFFFAOYSA-N |
| Synonym | 1,8-diazabicyclo 5.4.0 undec-7-ene,2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine,polycat dbu,dbu,unii-h1ilj6ibux,pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro,1,8-diazabicyclo 5.4.0-7-undecene,h1ilj6ibux,1,8-diazabicyclo 5,4,0 undec-7-ene |
| PubChem CID | 81184 |
| IUPAC Name | 2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine |
| SMILES | C1CCC2=NCCCN2CC1 |
Specification
| Chemical Name or Material | 1,8-Diazabicyclo[5.4.0]undec-7-ene |
|---|---|
| CAS | 6674-22-2 |
| Melting Point | -70°C |
| Density | 1.02 |
| pH | 12.8 |
| Boiling Point | 261°C |
| Flash Point | 116°C (240°F) |
| Odor | Amine-like |
| Molecular Formula | C9H16N2 |
| Refractive Index | 1.522 |
| MDL Number | MFCD00006930 |
| Quantity | 100 g |
| UN Number | UN3267 |
| Beilstein | 508906 |
| Sensitivity | Air Sensitive |
| Synonym | 1,8-diazabicyclo 5.4.0 undec-7-ene,2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine,polycat dbu,dbu,unii-h1ilj6ibux,pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro,1,8-diazabicyclo 5.4.0-7-undecene,h1ilj6ibux,1,8-diazabicyclo 5,4,0 undec-7-ene |
| Synonym | 1,8-diazabicyclo 5.4.0 undec-7-ene,2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine,polycat dbu,dbu,unii-h1ilj6ibux,pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro,1,8-diazabicyclo 5.4.0-7-undecene,h1ilj6ibux,1,8-diazabicyclo 5,4,0 undec-7-ene |
| Synonym | 1,8-diazabicyclo 5.4.0 undec-7-ene,2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine,polycat dbu,dbu,unii-h1ilj6ibux,pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro,1,8-diazabicyclo 5.4.0-7-undecene,h1ilj6ibux,1,8-diazabicyclo 5,4,0 undec-7-ene |
| Synonym | 1,8-diazabicyclo 5.4.0 undec-7-ene,2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine,polycat dbu,dbu,unii-h1ilj6ibux,pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro,1,8-diazabicyclo 5.4.0-7-undecene,h1ilj6ibux,1,8-diazabicyclo 5,4,0 undec-7-ene |
| Synonym | 1,8-diazabicyclo 5.4.0 undec-7-ene,2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine,polycat dbu,dbu,unii-h1ilj6ibux,pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro,1,8-diazabicyclo 5.4.0-7-undecene,h1ilj6ibux,1,8-diazabicyclo 5,4,0 undec-7-ene |
| Synonym | 1,8-diazabicyclo 5.4.0 undec-7-ene,2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine,polycat dbu,dbu,unii-h1ilj6ibux,pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro,1,8-diazabicyclo 5.4.0-7-undecene,h1ilj6ibux,1,8-diazabicyclo 5,4,0 undec-7-ene |
| Synonym | 1,8-diazabicyclo 5.4.0 undec-7-ene,2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine,polycat dbu,dbu,unii-h1ilj6ibux,pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro,1,8-diazabicyclo 5.4.0-7-undecene,h1ilj6ibux,1,8-diazabicyclo 5,4,0 undec-7-ene |
| Synonym | 1,8-diazabicyclo 5.4.0 undec-7-ene,2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine,polycat dbu,dbu,unii-h1ilj6ibux,pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro,1,8-diazabicyclo 5.4.0-7-undecene,h1ilj6ibux,1,8-diazabicyclo 5,4,0 undec-7-ene |
| Synonym | 1,8-diazabicyclo 5.4.0 undec-7-ene,2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine,polycat dbu,dbu,unii-h1ilj6ibux,pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro,1,8-diazabicyclo 5.4.0-7-undecene,h1ilj6ibux,1,8-diazabicyclo 5,4,0 undec-7-ene |
| Solubility Information | Miscible with water. |
| InChI Key | GQHTUMJGOHRCHB-UHFFFAOYSA-N |
| SMILES | C1CCC2=NCCCN2CC1 |
| IUPAC Name | 2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine |
| Molecular Weight (g/mol) | 152.241 |
| PubChem CID | 81184 |
| Formula Weight | 152.24 |
| Percent Purity | 99% |
Technical documents
[1,8-Diazabicyclo[5.4.0]undec-7-ene]
[Danger]
[{code=H301, sequence=1, description=Toxic if swallowed.}, {code=H314, sequence=2, description=Causes severe skin burns and eye damage.}, {code=H318, sequence=3, description=Causes serious eye damage.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=52, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=53, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=55, description=Immediately call a POISON CENTER/doctor/.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: 1,8-Diazabicyclo[5.4.0]undec-7-ene}]
[{code=GHS05, sequence=1, description=CORROSION}, {code=GHS06, sequence=2, description=SKULL & CROSSBONES}]
Product Identifier
- 1,8-Diazabicyclo[5.4.0]undec-7-ene
Signal Word
- Danger
Hazard Statement
- H301- Toxic if swallowed.
- H314- Causes severe skin burns and eye damage.
- H318- Causes serious eye damage.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
Supplemental Information
- MIXTURE LIST- Contain: 1,8-Diazabicyclo[5.4.0]undec-7-ene