Code:
85
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Standard
Supplement cell culture growth with U.S. sourced quality cell culture-grade Cytiva HyClone™ Standard Fetal Bovine Serum (FBS)
Additional Details:
Weight:
1.00000kg
Specification
Additional Assays | Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, IgG |
---|---|
Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
Description | Standard Fetal Bovine Serum, US Origin |
Country of Origin | United States |
Product Type | Fetal Bovine Serum |
Quantity | 500 mL |
Sterility | Sterile |
Sterilization Method | Sequential-filtered, 100 nm |
Grade | Standard |
Processing | None |
Tested For | Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |
Endotoxin Concentration | For Information Only |
Heat Inactivated | No |
Hemoglobin Concentration | For Information Only |
Mycoplasma Detected | No |
USDA Safety Testing Included | USDA Inspected |