Use promo code "24112" to get your promotional price.
Gibco™ Advanced MEM
Advanced MEM (Minimum Essential medium) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation.
15.26 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Use promo code "24112" to get your promotional price.
Code:
50
Additional Details:
Weight:
0.59000kg
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specification
Cell Line | HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts |
---|---|
Classification | Animal Origin |
Concentration | 1 X |
Form | Liquid |
Product Type | MEM (Minimum Essential Medium) |
Serum Level | Reduced Serum |
Sterility | Sterile-filtered |
Sterilization Method | Sterile-filtered |
With Additives | Low Glucose,Phenol Red,Sodium Pyruvate,Non Essential Amino Acids (NEAA) |
With Additives | Low Glucose,Phenol Red,Sodium Pyruvate,Non Essential Amino Acids (NEAA) |
With Additives | Low Glucose,Phenol Red,Sodium Pyruvate,Non Essential Amino Acids (NEAA) |
With Additives | Low Glucose,Phenol Red,Sodium Pyruvate,Non Essential Amino Acids (NEAA) |
Without Additives | No Glutamine,No HEPES |
Without Additives | No Glutamine,No HEPES |
Manufacturing Quality | cGMP-compliant under the ISO 13485 standard |
Product Line | Gibco™ |
Quantity | 500 mL |
Shipping Condition | Ambient |
Content And Storage |
Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |