73.33 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Use promo code "24112" to get your promotional price.
Content And Storage | Store in freezer (-5 to -20°C) and protect from light. |
---|---|
Cell Type | Neural Cells |
Classification | Serum-free |
Concentration | 50 X |
Culture Type | Mammalian Cell Culture |
Form | Liquid |
Product Type | Cell Culture Supplement |
Serum Level | Serum-Free |
Sterility | Sterile |
pH | 6 to 8 |
Product Line | B-27™ |
Quantity | 10 mL |
Shipping Condition | Dry Ice |
Source | Serum-Free |
Tested For | Endotoxin, Performance, Sterility, Mycoplasma |