Gibco™ Penicillin-Streptomycin (10,000 U/mL)
For prevention of bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria
Overview
7.64 GBP valid until 2025-08-31
Use promo code "24157" to get your promotional price.
Code: 50
Additional Details:
Additional Details: Weight: 0.03000kg
Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Product Code. 11568876
Quantity Price
1 £ 9.79 / 20mL
In stock, ready to ship: 55
Add to basket
For Research Use Only. All usage must comply with product instructions.
  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
Description and Specification
Specification
Content And Storage Storage conditions: -5 to -20°C
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture
Concentration 100 X
Form Liquid
Product Type Antibiotic
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
For Use With (Application) Prevention of Cell Culture Contamination
Quantity 20 mL
Shipping Condition Dry Ice
Penicillin was originally purified from fungi Penicillium that acts by interfering directly with turnover of bacterial cell wall and indirectly by triggering release of enzymes that alter the cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to 30S subunit of bacterial ribosome, leading to inhibition of protein synthesis and death in susceptible bacteria.
  • Contains 10,000 units/mL of penicillin
  • 10,000μg/mL of streptomycin in a 10 mM citrate buffer (for pH stability)
Alternative Products
Documentation
Technical documents
Safety and Handling
[Penicillin-Streptomycin, liquid]
[Warning]
[{code=H361, sequence=1, description=Suspected of damaging fertility or the unborn child.}]
[{code=P201, sequence=51, description=Obtain special instructions before use.}, {code=P202, sequence=52, description=Do not handle until all safety precautions have been read and understood.}, {code=P280, sequence=53, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P308+P313, sequence=54, description=IF exposed or concerned: Get medical advice/attention.}, {code=P501b, sequence=55, description=Dispose of contents/container in accordance with local/regional/national/international regulations.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: streptomycin sulphate}]
[{code=GHS08, sequence=1, description=HEALTH HAZARD}]
Product Identifier
  • Penicillin-Streptomycin, liquid
Signal Word
  • Warning
Hazard Statement
  • H361- Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201- Obtain special instructions before use.
  • P202- Do not handle until all safety precautions have been read and understood.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313- IF exposed or concerned: Get medical advice/attention.
  • P501b- Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental Information
  • MIXTURE LIST- Contain: streptomycin sulphate