Use promo code "24335" to get your promotional price.
Gibco™ Trypsin-EDTA (0.25%), phenol red
Use promo code "24335" to get your promotional price.
Specification
| Chelators | EDTA |
|---|---|
| Content And Storage |
Storage conditions: -5°C to -20°C Shipping conditions: Wet ice Shelf life: 24 months from date of manufacture |
| Cell Type | Mammalian Cells |
| Classification | Animal Origin |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | Cell Culture Dissociation Reagent |
| Sterility | Sterile-filtered |
| Sterilization Method | Sterile-filtered |
| With Additives | Phenol Red,EDTA |
| With Additives | Phenol Red,EDTA |
| Osmolality | 270 - 320 mOsm/kg |
| pH | 7.1 to 8 |
| Product Line | Gibco |
| Quantity | 500 mL |
| Shipping Condition | Wet Ice |
| Source | Animal Origin |
| Tested For | In vitro Bioassay |
Technical documents
[Trypsin, 0.25% (1X) with EDTA 4Na, liquid]
[Danger]
[{code=H334, sequence=1, description=May cause allergy or asthma symptoms or breathing difficulties if inhaled.}]
[{code=P261, sequence=51, description=Avoid breathing dust/fume/gas/mist/vapours/spray.}, {code=P284, sequence=52, description=¢In case of inadequate ventilation! wear respiratory protection.}, {code=P304+P340, sequence=53, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P342+P311, sequence=54, description=If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .}, {code=P501b, sequence=55, description=Dispose of contents/container in accordance with local/regional/national/international regulations.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: trypsin}]
[{code=GHS08, sequence=1, description=HEALTH HAZARD}]
Product Identifier
- Trypsin, 0.25% (1X) with EDTA 4Na, liquid
Signal Word
- Danger
Hazard Statement
- H334- May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261- Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284- ¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311- If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b- Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental Information
- MIXTURE LIST- Contain: trypsin