Code:
NEW
Gibco™ DMEM, high glucose
DMEM, high glucose
Additional Details:
Weight:
5.00000kg
Specification
Concentration | 1 X |
---|---|
Form | Liquid |
Product Type | Dulbecco's Modified Eagle Medium |
With Additives | High Glucose,Glutamine,Phenol Red |
With Additives | High Glucose,Glutamine,Phenol Red |
With Additives | High Glucose,Glutamine,Phenol Red |
Without Additives | No HEPES,No Sodium Pyruvate |
Without Additives | No HEPES,No Sodium Pyruvate |
For Use With (Application) | Tissue and cell culture processing, mammalian cell culture |
Quantity | 10 x 500mL |
Certifications/Compliance | ISO 13485 standards, FDA |
pH | 7.0 to 7.4 |
Content And Storage | 2 to 8°C |
Description | Dulbecco's Modified Eagle Medium (DMEM), 10 x 500mL, Liquid, 1 X Concentration, 7.0 to 7.4 pH, With High Glucose, L-glutamine, Phenol Red, 12 Months Shelf Life, 320 to 360mOsm/kg Osmolality, For Mammalian Cell Culture |