Zwiebel™ Class F1 Box Weights
Use with weighing instruments of medium accuracy class II. Zwiebel™ Class F1 Box Weights are ideal for the verification or calibration of class F2 weights.
Overview
Code: A0
Additional Details:
Additional Details: Weight: 0.80000kg
Product Code. 11730703
Quantity Price
1 £ 58.8 / Each
Estimated Shipment
03-06-2026
Add to basket
  • Description and Specification
  • Alternative Products
  • Accessories and Spare Parts
Description and Specification
Specification
Mass 100 g
Tolerance ±0.50 mg
For Use With (Application) For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Alternative Products
Accessories and Spare Parts