Code:
50
Memmert™ Vacuum Oven Thermoshelf
Additional Details:
Weight:
1.00000kg
Specification
Certifications/Compliance | Calibration certificate |
---|---|
For Use With (Equipment) | Model 200 |
Type | Shelf |
For Use With (Application) | Individual overtemperature protection for each shelf MLOP (Multi-Level-Overtemperature-Control) |
Includes | Local Temperature Sensing (Pt100, 4-Wire-Circuit) |
Material | Stainless Steel |