541.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
| Quantity | 2 L |
|---|---|
| Packaging | LDPE Bottle |
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.015 |
| MDL Number | 11332 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | dihydrogen oxide,dihydrogen monoxide |
| PubChem CID | 962 |
| ChEBI | CHEBI:15377 |
| IUPAC Name | oxidane |
| SMILES | O |
| Chemical Name or Material | Water |
|---|---|
| pH | 7 |
| CAS | 7732-18-5 |
| Color | Colorless |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Molecular Formula | H2O |
| MDL Number | 11332 |
| Synonym | dihydrogen oxide,dihydrogen monoxide |
| Synonym | dihydrogen oxide,dihydrogen monoxide |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| SMILES | O |
| IUPAC Name | oxidane |
| Molecular Weight (g/mol) | 18.015 |
| PubChem CID | 962 |
| ChEBI | CHEBI:15377 |
| Formula Weight | 18.02 |
| Grade | Ultra Trace Elemental Analysis |
| Packaging | LDPE Bottle |
| Vapor Pressure | 17.5mmHg at 20°C |
| Density | 1.000g/cm³ |
| Certificate of Analysis | Certificate of Lot Analysis included with each bottle. |
| Quantity | 2 L |
| Physical Form | Liquid |