Thermo Scientific Chromacol Chromacol Nylon Syringe Filters
Thermo Scientific™ Chromacol Nylon Syringe Filters are available in a selection of diameters and pore sizes.
Overview
147.30 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: 17
Additional Details:
Additional Details: Weight: 0.50000kg
Product Code. 12304787
Quantity Price
1 £ 190.0 / Pack of 100
Estimated Shipment
26-08-2025
Add to basket
  • Description and Specification
  • Alternative Products
Description and Specification
Specification
Description 30-SF-45(N)P
Material (Membrane) Nylon
Pore Size 0.45 μm
Prefilter Yes
Type Syringe Filter
Diameter (Metric) 30 mm
Alternative Products