44.20 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Use promo code "24155" to get your promotional price.
Capacity (Metric) | 500 mL |
---|---|
Capacity (English) | 16 oz. |
Color | Translucent,Blue |
Color | Translucent,Blue |
Includes | Pack of 4 each of “DI Water” wash botttles |
Material | LDPE |
Quantity | 4/Pk. |
Sterility | Non-sterile |
Closure Color | Blue |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Disposable | Reusable |
Graduated | No |
Mouth | Wide, Threaded |
Shape | Cylindrical |
Product Type | Wash Bottle |
Chemical Label | Deionized Water |
Closure Material | Polypropylene |