1037.51 GBP valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Use promo code "24075" to get your promotional price.
Certifications/Compliance | CE, cUL, DVE, EAC, GS |
---|---|
Chamber Material | Stainless Steel (304/1.4301) |
Controller Type | SingleDISPLAY. Adaptive multifunctional digital PID-microprocessor controller |
Convection Type | Natural |
Capacity (Metric) | 53 L |
Temperature (Metric) | 20°C to 300°C |
Type | Heating Oven |
Display | 1 high-definition TFT-colour display (SingleDISPLAY) |
For Use With (Application) | Temperature Control in Science, Research and Material Tests in Industry |
Includes | PC Software AtmoCONTROL |
Insulation | Fully isolated Stainless Steel door with 2-point locking (compression door lock) |
Material | Stainless Steel |
Max. Shelf Load | 20 kg |
Model | UN55 |
No. of Shelves | 4 |
Plug Type | Mains cable with plug (German type) |
Safety Features | Temperature control, autodiagnostic system |
Shelf Material | Stainless Steel |
Temperature Control | Adaptive multifunctional digital PID-microprocessor controller |
Temperature Stability | Up to 99.9°C: 0.1 / from 100°C: 0.5 |
Timer | Digital backwards counter with target time setting, adjustable from 1 minute to 99 days |
Warranty | 3 years |
Dimensions (D x W x H) Interior | 330 x 400 x 400 mm |
Dimensions (L x W x H) | 585 x 784 x 514 mm |
Length (Metric) | 514 mm |
Width (Metric) | 585 mm |
Width (Metric) Interior | 400 mm |
Height (Metric) | 784 mm |
Height (Metric) Interior | 400 mm |
Depth (Metric) | 514 mm |
Depth (Metric) Interior | 330 mm |
Shipping Weight (Metric) | 67 kg |
Weight (Metric) | 57 kg |
Electrical Requirements | 115/230 V, 50/60 Hz |
Voltage | 115/230 V |
Wattage | 1700/2000 W |
Frequency | 50/60 Hz |