Syngene™ Cy5 Filter
Syngene™ Cy5 Filter works in conjuction with the Syngene Gbox and Gbox Mini Systems.
Overview
Code: 04
Additional Details:
Additional Details: Weight: 0.50000kg
Product Code. 12843998
Quantity Price
1 £ 7445.0 / Each
Estimated Shipment
05-08-2025
Add to basket
Description and Specification
Specification
For Use With (Equipment) Gbox and Gbox Mini
Wavelength 692nm
  • For use in GBOX and Gbox Mini systems 7 position filter wheel.