Use promo code "24117" to get your promotional price.
Thermo Scientific™ GeneJET RNA Cleanup and Concentration Micro Kit
Use promo code "24117" to get your promotional price.
Specification
Content And Storage | For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C. |
---|---|
Format | Spin Column |
Isolation Technology | Spin Column |
Purification Time | 5 min. |
Sample Type | RNA |
High-throughput Compatibility | Not High-throughput Compatible (Manual) |
For Use With (Application) | Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Nuclease Protection Assays, Nucleic Acid Labeling, Northern Blotting, cDNA Library Construction |
No. of Reactions | 50 Preps |
Product Line | GeneJET |
Quantity | 50 Preps |
Scale | Micro |
Target | Total RNA |
[GeneJET RNA Cleanup and Concentration Micro Kit]
[Danger]
[{code=H302, sequence=1, description=Harmful if swallowed.}, {code=H314, sequence=2, description=Causes severe skin burns and eye damage.}, {code=H315, sequence=3, description=Causes skin irritation.}, {code=H319, sequence=4, description=Causes serious eye irritation.}, {code=H412, sequence=5, description=Harmful to aquatic life with long lasting effects.}]
[{code=P260, sequence=51, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P264, sequence=52, description=Wash hands thoroughly after handling.}, {code=P270, sequence=53, description=Do not eat, drink or smoke when using this product.}, {code=P273, sequence=54, description=Avoid release to the environment.}, {code=P280, sequence=55, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P310, sequence=56, description=IF SWALLOWED: Immediately call a POISON CENTER/doctor /}, {code=P301+P312, sequence=57, description=IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.}, {code=P301+P330+P331, sequence=58, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P302+P352, sequence=59, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P303+P361+P353, sequence=60, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P304+P340, sequence=61, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=62, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=63, description=Immediately call a POISON CENTER/doctor/.}, {code=P330, sequence=64, description=Rinse mouth.}, {code=P332+P313, sequence=65, description=If skin irritation occurs: Get medical advice/attention.}, {code=P363, sequence=66, description=Wash contaminated clothing before reuse.}, {code=P501b, sequence=67, description=Dispose of contents/container in accordance with local/regional/national/international regulations.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Thiocyanic acid, Guanidine hydrochloride}]
[{code=GHS05, sequence=1, description=CORROSION}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- GeneJET RNA Cleanup and Concentration Micro Kit
Signal Word
- Danger
Hazard Statement
- H302- Harmful if swallowed.
- H314- Causes severe skin burns and eye damage.
- H315- Causes skin irritation.
- H319- Causes serious eye irritation.
- H412- Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P260- Do not breathe dust/fume/gas/mist/vapours/spray.
- P264- Wash hands thoroughly after handling.
- P270- Do not eat, drink or smoke when using this product.
- P273- Avoid release to the environment.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310- IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312- IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352- IF ON SKIN: Wash with plenty of water/soap.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
- P330- Rinse mouth.
- P332+P313- If skin irritation occurs: Get medical advice/attention.
- P363- Wash contaminated clothing before reuse.
- P501b- Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental Information
- MIXTURE LIST- Contains : Thiocyanic acid, Guanidine hydrochloride