HUBER CC-K6 Refrigerated Heating Bath
An economic solution for controlling the temperature of objects directly within the thermostat bath. The units are suitable for a number of typical heating and cooling tasks such as sample temperature control, materials testing, quality control, and analyses
Overview
4970.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: A0
Additional Details:
Additional Details: Weight: 25.00000kg
Product Code. 13361689
Quantity Price
1 £ 6865.0 / Each
Estimated Shipment
07-07-2026
Add to basket
  • Description and Specification
  • Alternative Products
Description and Specification
Specification
Cooling Capacity 20°C at 0.2 kW, 0°C at 0.15 kW, -10°C at 0.1°CkW, to 20°C (at 0.05 kW)
Pump Type Circulation
Temperature Range -25°C to +200°C
Display TFT
Features TFT Touchscreen Display , E-grade Fuction, Integrated Programmer, USB Inferance, Ethernet Inferance, RS232 inferance, Plug and play control technology, Compatible with spycontrol,Easy control operating concept, Protection safety fuction, Natural refrigerant
Heating Capacity 1.6 - 2.1 kW
Model CC-K6
Pump Capacity 27 L / min; 0.7 bar Pressure, 22 L / min; 0.4 bar Capacity
Safety Class 3/FL
Temperature Stability ±0.02°C
Warranty 3 years for removable (Plug & Play) electronic controls, 2 years for refrigeration components, 1 year for mechanical and electrical components
Width (Metric) 210 mm
Height (Metric) 546 mm
Weight (Metric) 25 kg
Voltage 208/240 V
Frequency 50/60 Hz
  • Refrigerated Heating Bath
  • Temperature range: -25°C to 200°C
  • Cooling capacity (20°C): 0.2kW
  • Cooling capacity (0°C): 0.15kW
  • Heating capacity: 1,6 - 2,1 kW
  • Pressure pump max.: 27L/min / 0.7bar
  • Suction pump max.: 22L/min / 0.4bar
  • Pump connection: M16 x 1 male
  • Bath volume: 4.5L
  • Power supply: 208-240V 1∼/ 2∼ 50/60Hz. With natural refrigerant.
Alternative Products