150.28 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
| Capacity (Metric) | 500 mL |
|---|---|
| For Use With (Application) | Storage of air sensitive reagents, and chemicals; Storage of hygroscopic liquids, powders and granules |
| Includes | With standard ground glass flat-headed stopper |
| Material | Soda Lime Glass |
| Quantity | 10/Pk. |
| Diameter (Metric) | 89 mm |
| Autoclavable | Not Autoclavable |