| Chemical Name or Material |
Sodium thiosulfate
|
| Melting Point |
48.0°C
|
| CAS |
7772-98-7
|
| pH |
6.0 to 8.4 (5% aq. soln.)
|
| Quantity |
1 kg
|
| Assay Percent Range |
98.5% min. (Iodimetry)
|
| Molecular Formula |
Na2O3S2
|
| Linear Formula |
Na2S2O3
|
| MDL Number |
MFCD00003499
|
| Merck Index |
15,8821
|
| Synonym |
sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
|
| Synonym |
sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
|
| Synonym |
sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
|
| Synonym |
sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
|
| Synonym |
sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
|
| Synonym |
sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
|
| Synonym |
sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
|
| Synonym |
sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
|
| Synonym |
sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
|
| Synonym |
sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt
|
| Solubility Information |
Solubility in water: 700g/L (20°C). Other solubilities: practically insoluble in alcohol
|
| InChI Key |
AKHNMLFCWUSKQB-UHFFFAOYSA-L
|
| SMILES |
[Na+].[Na+].[O-]S([S-])(=O)=O
|
| IUPAC Name |
disodium sulfanidesulfonate
|
| Molecular Weight (g/mol) |
158.10
|
| PubChem CID |
24477
|
| Formula Weight |
158.1
|
| Percent Purity |
99%
|
| Grade |
Analytical
|
| Loss on Drying |
1% max. (105°C)
|