Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol
CAS: 124-41-4 | CH3NaO | 54.02 g/mol
Overview
404.52 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: CAS Number: 124-41-4 Weight: 24.25000kg Transport: UN number : 1289 Chem class : 3 Pack group : III
Quantity 25 L
Disclaimers: RUO – Research Use Only
Product Code. 15294654
Quantity Price
1 £ 531.0 / 25 L
Estimated Shipment
03-07-2026
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
Description and Specification
Chemical Identifiers
CAS 124-41-4
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.02
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]
Specification
Chemical Name or Material Sodium methoxide
Name Note 30 wt% Solution in Methanol
CAS 124-41-4
CAS 67-56-1
Melting Point 1.0°C to 5.0°C
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 93.0°C
Flash Point 33°C
Molecular Formula CH3NaO
Linear Formula CH3ONa
Refractive Index 1.3701
MDL Number MFCD00012179
Quantity 25 L
Beilstein 01,28
Fieser 01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352
Merck Index 15,8775
Specific Gravity 0.97
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Solubility Information Solubility in water: reacts. Other solubilities: soluble in methanol
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
SMILES C[O-].[Na+]
IUPAC Name sodium;methanolate
Molecular Weight (g/mol) 54.02
PubChem CID 10942334
Viscosity 68 mPa.s (20°C)
Formula Weight 54.02
Grade Pure
Physical Form Liquid
Alternative Products
Documentation
Technical documents
Safety and Handling
[Sodium methoxide, 5.4M (30 wt.%) solution in methanol]
[Danger]
[{code=H226, sequence=1, description=Flammable liquid and vapour.}, {code=H290, sequence=2, description=May be corrosive to metals.}, {code=H301, sequence=3, description=Toxic if swallowed.}, {code=H311, sequence=4, description=Toxic in contact with skin.}, {code=H314, sequence=5, description=Causes severe skin burns and eye damage.}, {code=H318, sequence=6, description=Causes serious eye damage.}, {code=H331, sequence=7, description=Toxic if inhaled.}, {code=H370, sequence=8, description=Causes damage to organs.}, {code=EUH014, sequence=91, description=Reacts violently with water.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=53, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=54, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P308+P313, sequence=56, description=IF exposed or concerned: Get medical advice/attention.}, {code=P310, sequence=57, description=Immediately call a POISON CENTER/doctor/.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: Methanol, Sodium methylate}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS05, sequence=2, description=CORROSION}, {code=GHS06, sequence=3, description=SKULL & CROSSBONES}, {code=GHS08, sequence=4, description=HEALTH HAZARD}]
Product Identifier
  • Sodium methoxide, 5.4M (30 wt.%) solution in methanol
Signal Word
  • Danger
Hazard Statement
  • H226- Flammable liquid and vapour.
  • H290- May be corrosive to metals.
  • H301- Toxic if swallowed.
  • H311- Toxic in contact with skin.
  • H314- Causes severe skin burns and eye damage.
  • H318- Causes serious eye damage.
  • H331- Toxic if inhaled.
  • H370- Causes damage to organs.
  • EUH014- Reacts violently with water.
Precautionary Statement
  • P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313- IF exposed or concerned: Get medical advice/attention.
  • P310- Immediately call a POISON CENTER/doctor/.
Supplemental Information
  • MIXTURE LIST- Contain: Methanol, Sodium methylate