Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications, Corning™
High quality water for use in cell culture applications. Corning™ Cell Culture Grade Water Tested to USP Sterile Water are used in preparation of purification and chromatography buffers.
Overview
114.19 GBP valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: CAS Number: 7732-18-5 Weight: 6.98000kg
Quantity 6 x 1 L
Packaging Bottle
Alerts: Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
Product Code. 15373581
Quantity Price
1 £ 161.71 / Pack of 6
In stock, ready to ship: 100
Add to basket
  • Description and Specification
  • Alternative Products
Description and Specification
Specification
CAS 7732-18-5
Packaging Bottle
Quantity 6 x 1 L
Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).
Alternative Products