Use promo code "27189" to get your promotional price.
Invitrogen™ Novex™ Tris-Glycine Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format
Invitrogen Novex Tris-Glycine Mini Gels, WedgeWell format, are polyacrylamide gels that provide reproducible separation of a wide range of proteins into well-resolved bands with two times the loading capacity.
72.42 GBP valid until 2025-12-31
Use promo code "27189" to get your promotional price.
Use promo code "27189" to get your promotional price.
Code:
NEW
Additional Details:
Weight:
1.07000kg
Specification
| Gel Percentage | 10% |
|---|---|
| Gel Size | Mini |
| GelThickness | 1.0 mm |
| Gel Type | Tris-Glycine Plus |
| Recommended Applications | Denaturing, Native |
| Sample Loading Volume | Up to 45 μL |
| Separation Range | 30 to 250 kDa |
| Separation Type | Denaturing, Native |
| Wells | 12-well |
| Mode of Separation | Molecular Weight |
| Quantity | 10 gels (1 box) |
| Shipping Condition | Wet Ice |
| Storage Requirements | Store at 2°C to 8°C. Do not freeze. |
| Length (Metric) | 8 cm |
| Width (Metric) | 8 cm |
| Product Line | Novex, WedgeWell |
| For Use With (Equipment) | XCell SureLock Mini-Cell |