Chemical Name or Material |
odium perchlorate monohydrate
|
Melting Point |
130°C (decomposition)
|
Name Note |
For analysis ACS
|
CAS |
7791-07-3
|
Color |
White
|
Physical Form |
Crystalline
|
Quantity |
1 kg
|
Assay Percent Range |
85.0-90.0%
|
Molecular Formula |
ClH2NaO5
|
MDL Number |
MFCD00149164
|
Synonym |
sodium perchlorate monohydrate,sodium perchlorate hydrate,unii-g4h485lhcw,g4h485lhcw,ccris 8893,perchloric acid, sodium salt, monohydrate,sodium hydrate perchlorate,clo4.na.h2o,acmc-1bii1,sodium perchlorate-monohydrate
|
Synonym |
sodium perchlorate monohydrate,sodium perchlorate hydrate,unii-g4h485lhcw,g4h485lhcw,ccris 8893,perchloric acid, sodium salt, monohydrate,sodium hydrate perchlorate,clo4.na.h2o,acmc-1bii1,sodium perchlorate-monohydrate
|
Synonym |
sodium perchlorate monohydrate,sodium perchlorate hydrate,unii-g4h485lhcw,g4h485lhcw,ccris 8893,perchloric acid, sodium salt, monohydrate,sodium hydrate perchlorate,clo4.na.h2o,acmc-1bii1,sodium perchlorate-monohydrate
|
Synonym |
sodium perchlorate monohydrate,sodium perchlorate hydrate,unii-g4h485lhcw,g4h485lhcw,ccris 8893,perchloric acid, sodium salt, monohydrate,sodium hydrate perchlorate,clo4.na.h2o,acmc-1bii1,sodium perchlorate-monohydrate
|
Synonym |
sodium perchlorate monohydrate,sodium perchlorate hydrate,unii-g4h485lhcw,g4h485lhcw,ccris 8893,perchloric acid, sodium salt, monohydrate,sodium hydrate perchlorate,clo4.na.h2o,acmc-1bii1,sodium perchlorate-monohydrate
|
Synonym |
sodium perchlorate monohydrate,sodium perchlorate hydrate,unii-g4h485lhcw,g4h485lhcw,ccris 8893,perchloric acid, sodium salt, monohydrate,sodium hydrate perchlorate,clo4.na.h2o,acmc-1bii1,sodium perchlorate-monohydrate
|
Synonym |
sodium perchlorate monohydrate,sodium perchlorate hydrate,unii-g4h485lhcw,g4h485lhcw,ccris 8893,perchloric acid, sodium salt, monohydrate,sodium hydrate perchlorate,clo4.na.h2o,acmc-1bii1,sodium perchlorate-monohydrate
|
Synonym |
sodium perchlorate monohydrate,sodium perchlorate hydrate,unii-g4h485lhcw,g4h485lhcw,ccris 8893,perchloric acid, sodium salt, monohydrate,sodium hydrate perchlorate,clo4.na.h2o,acmc-1bii1,sodium perchlorate-monohydrate
|
Synonym |
sodium perchlorate monohydrate,sodium perchlorate hydrate,unii-g4h485lhcw,g4h485lhcw,ccris 8893,perchloric acid, sodium salt, monohydrate,sodium hydrate perchlorate,clo4.na.h2o,acmc-1bii1,sodium perchlorate-monohydrate
|
Synonym |
sodium perchlorate monohydrate,sodium perchlorate hydrate,unii-g4h485lhcw,g4h485lhcw,ccris 8893,perchloric acid, sodium salt, monohydrate,sodium hydrate perchlorate,clo4.na.h2o,acmc-1bii1,sodium perchlorate-monohydrate
|
Solubility Information |
Soluble in water.
|
InChI Key |
IXGNPUSUVRTQGW-UHFFFAOYSA-M
|
SMILES |
O.[O-]Cl(=O)(=O)=O.[Na+]
|
IUPAC Name |
sodium;perchlorate;hydrate
|
Molecular Weight (g/mol) |
140.45
|
PubChem CID |
516899
|
Formula Weight |
140.48 (122.46 Anhydrous)
|
Grade |
ACS
|
Sensitivity |
Hygroscopic
|
Appearance |
White crystals
|
Density |
2.02 g/cm3 at 20°C
|