13.19 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Use promo code "24127" to get your promotional price.
Capacity | 3.3 oz., 100 mL |
---|---|
Capacity (Metric) | 100 mL |
Capacity (English) | 3.3 oz. |
Color | Natural |
Diameter (English) | 2.2 in. |
Diameter (English) Inner Neck | 1.18 in. |
Diameter (Metric) Inner Neck | 30 mm |
Includes | Clear drip-free Polypropylene pouring ring |
Material | Glass |
Sterility | Non-sterile |
Closure Color | Blue |
Graduations | 20 to 80mL |
Closure Included | Yes |
Autoclavable | Autoclavable |
Graduated | Yes |
Mouth | Wide |
Shape | Round |
Height (Metric) | 100 mm |
Product Type | Media Bottle |
Height (English) | 3.9 in. |
Closure Material | Polypropylene |
Closure Size | GL 45 |
Diameter (Metric) | 56 mm |
Closure Type | Screw Cap |