Code:
NEW
Petroleum ether, ACS reagent, Honeywell Riedel-de Haën™
ACS reagent
Additional Details:
Weight:
3.90000kg
Transport:
UN number : 3295
Chem class : 3
Pack group : II
| Quantity | 6 x 1 L |
|---|---|
| Packaging | Glass Bottle |
Chemical Identifiers
| CAS | 8030-30-6 |
|---|---|
| Molecular Formula | C5H12 |
| Molecular Weight (g/mol) | 72.15 |
| MDL Number | MFCD00009498 |
| InChI Key | OFBQJSOFQDEBGM-UHFFFAOYSA-N |
| Synonym | n-pentane,pentan,skellysolve a,pentanen,pentani,amyl hydride,tetrafume,tetrakil,tetraspot,pentan polish |
| PubChem CID | 8003 |
| ChEBI | CHEBI:37830 |
| IUPAC Name | pentane |
| SMILES | CCCCC |
Specification
| Chemical Name or Material | Petroleum ether |
|---|---|
| Name Note | ACS reagent |
| CAS | 8030-30-6 |
| CAS Min % | ≥25.0000% |
| CAS Max % | <50.0000% |
| Density | 0.64 g/mL at 25 °C(lit.) |
| Boiling Point | 40°C to 60°C |
| Flash Point | −30°C |
| Packaging | Glass Bottle |
| Molecular Formula | C5H12 |
| MDL Number | MFCD00009498 |
| Quantity | 6 x 1 L |
| UN Number | UN3295 |
| Synonym | n-pentane,pentan,skellysolve a,pentanen,pentani,amyl hydride,tetrafume,tetrakil,tetraspot,pentan polish |
| Synonym | n-pentane,pentan,skellysolve a,pentanen,pentani,amyl hydride,tetrafume,tetrakil,tetraspot,pentan polish |
| Synonym | n-pentane,pentan,skellysolve a,pentanen,pentani,amyl hydride,tetrafume,tetrakil,tetraspot,pentan polish |
| Synonym | n-pentane,pentan,skellysolve a,pentanen,pentani,amyl hydride,tetrafume,tetrakil,tetraspot,pentan polish |
| Synonym | n-pentane,pentan,skellysolve a,pentanen,pentani,amyl hydride,tetrafume,tetrakil,tetraspot,pentan polish |
| Synonym | n-pentane,pentan,skellysolve a,pentanen,pentani,amyl hydride,tetrafume,tetrakil,tetraspot,pentan polish |
| Synonym | n-pentane,pentan,skellysolve a,pentanen,pentani,amyl hydride,tetrafume,tetrakil,tetraspot,pentan polish |
| Synonym | n-pentane,pentan,skellysolve a,pentanen,pentani,amyl hydride,tetrafume,tetrakil,tetraspot,pentan polish |
| Synonym | n-pentane,pentan,skellysolve a,pentanen,pentani,amyl hydride,tetrafume,tetrakil,tetraspot,pentan polish |
| Synonym | n-pentane,pentan,skellysolve a,pentanen,pentani,amyl hydride,tetrafume,tetrakil,tetraspot,pentan polish |
| InChI Key | OFBQJSOFQDEBGM-UHFFFAOYSA-N |
| SMILES | CCCCC |
| IUPAC Name | pentane |
| Molecular Weight (g/mol) | 72.15 |
| PubChem CID | 8003 |
| ChEBI | CHEBI:37830 |
| Grade | ACS Reagent |