53.20 GBP valid until 2025-12-31
Use promo code "24071" to get your promotional price.
Use promo code "24071" to get your promotional price.
| Concentration | 1 μg/μL |
|---|---|
| Components | TrueCut Cas9 Protein v2 |
| Grade | Molecular Biology |
| Content And Storage |
TrueCut Cas9 Protein v2 (1 mg/mL), 10 μg Store at -5°C to -30°C. |
| Accession Number | NC_002737.2:854751-858857 |
| Expression System | E. coli |
| Gene ID (Entrez) | 901176 |
| Gene Symbol | Spy_1046 |
| Gene Alias | cas5, csn1 |
| Protein Form | Recombinant |
| Protein Tag | Untagged |
| Shipping Condition | Dry Ice |
| Validated Application | Genome Editing |
| Quantity | 10 μg |
| Product Type | Cas9 Protein v2 |
| Research Category | Genome Editing |
| Product Line | TrueCut |