Microspec™ Plastic Culture Needles
Transfer and inoculate living microorganisms
Additional Details:
Additional Details: Weight: 1.20000kg
Product Code. 15782125
Quantity Price
1 £ 55.85 / Pack of 1000
In stock, ready to ship: 4
Add to basket
  • Description and Specification
  • Alternative Products
Description and Specification
Specification
Disposable Single-use
Length (English) 7.9 in.
Length (Metric) 200 mm
Material High-impact Polystyrene
Sterility Aseptically Manufactured
Color Blue
Product Type Inoculating Needle
Quantity 1000
Packaging Type Packaged in Microseal Pouches of 20
Autoclavable Not Autoclavable
  • Sharp end to facilitate easy stab inoculation
  • Picks up small colonies
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured
Alternative Products